Learn More
Aviva Systems Biology Corp ST6GALNAC2 Antibody - middle region (ARP53653_P050) | 100uL
SDP
Catalog No. 5001461080
Encompass_Preferred
This is a rabbit polyclonal antibody against ST6GALNAC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST6GALNAC2. Alternate names: SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM, SAITL1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61080 Supplier Aviva Systems Biology Corp Supplier No. ARP53653_P050
This is a rabbit polyclonal antibody against ST6GALNAC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST6GALNAC2. Alternate names: SIAT7, SIAT7B, SIATL1, ST6GalNAII, STHM, SAITL1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat