Learn More
Aviva Systems Biology Corp ST6GALNAC3 Antibody - C-terminal region (ARP49075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST6GALNAC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ST6GALNAC3. Alternate names: PRO7177, SIAT7C, ST6GALNACIII, STYApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-60013