Learn More
Aviva Systems Biology Corp ST6GALNAC4 Antibody - middle region (ARP49121_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49121_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST6GALNAC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST6GALNAC4. Alternate names: SIAT3C, SIAT7D, ST6GALNACIVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53574