Learn More
Aviva Systems Biology Corp ST6GALNAC6 Antibody - N-terminal region (ARP46951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST6GALNAC6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ST6GALNAC6. Alternate names: RP11-203J24.3, SIAT7F, ST6GALNACVIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-277