Learn More
Aviva Systems Biology Corp ST8SIA2 Antibody - C-terminal region (ARP46681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST8SIA2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ST8SIA2. Alternate names: HsT19690, MGC116854, MGC116857, SIAT8B, ST8SIA-II, STXApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59376