Learn More
Aviva Systems Biology Corp ST8SIA4 Antibody - middle region (ARP48679_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48679_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST8SIA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST8SIA4. Alternate names: MGC34450, MGC61459, PST, PST1, SIAT8D, ST8SIA-IVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59883