Learn More
Aviva Systems Biology Corp ST8SIA6 Antibody - middle region (ARP44486_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44486_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ST8SIA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST8SIA6. Alternate names: SIAT8F, ST8SIA-VI, ST8Sia VI, SIA8FApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58825