Learn More
Apexbio Technology LLC STA-21 111540-00-2 1mg
SDP
Catalog No. 5000088092
Encompass_Preferred
STA-21 (CAS 111540-00-2) is a small-molecule inhibitor targeting STAT3 a transcription factor activated by cytokines such as IL-6 It inhibits STAT3 DNA-binding activity and dimerization thereby modulating downstream signaling pathways associated with Th17 differentiation and inflammatory processes STA-21 exerts its biological activity primarily by suppressing STAT3-mediated IL-17 expression and osteoclast differentiation as well as promoting FoxP3 expression In in vitro studies STA-21 inhibits DU145 cell viability with an IC50 of 12 2 M Based on these pharmacological properties STA-21 holds research potential in the fields of autoimmune and inflammatory disease
Catalog No. 50-000-88092 Supplier Apexbio Technology LLC Supplier No. B61751
STA-21 (CAS 111540-00-2) is a small-molecule inhibitor targeting STAT3 a transcription factor activated by cytokines such as IL-6 It inhibits STAT3 DNA-binding activity and dimerization thereby modulating downstream signaling pathways associated with Th17 differentiation and inflammatory processes STA-21 exerts its biological activity primarily by suppressing STAT3-mediated IL-17 expression and osteoclast differentiation as well as promoting FoxP3 expression In in vitro studies STA-21 inhibits DU145 cell viability with an IC50 of 12 2 M Based on these pharmacological properties STA-21 holds research potential in the fields of autoimmune and inflammatory disease