Learn More
Enzo Life Sciences STA-21, (1mg), CAS Number: 28882-53-3
SDP
Catalog No. NC9876078
Encompass_Preferred
STAT inhibitor. STA-21 is a novel Stat3-selective inhibitor. It inhibits Stat3 dimerization and DNA binding as well as Stat3-dependent luciferase reporter activity. Alternative Name: (S)-Ochromycinone, Deoxytetrangomycin. Formula: C19H14O4. MW: 306.3. CAS Number: 28882-53-3. Purity: ≥98% (HPLC). Appearance: White to yellow solid. Solubility: Soluble in DMSO (20mg/ml). Use/Stability: Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC9876078 Supplier Enzo Life Sciences Supplier No. BMLEI3510001
STAT inhibitor. STA-21 is a novel Stat3-selective inhibitor. It inhibits Stat3 dimerization and DNA binding as well as Stat3-dependent luciferase reporter activity. Alternative Name: (S)-Ochromycinone, Deoxytetrangomycin. Formula: C19H14O4. MW: 306.3. CAS Number: 28882-53-3. Purity: ≥98% (HPLC). Appearance: White to yellow solid. Solubility: Soluble in DMSO (20mg/ml). Use/Stability: Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C