Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-(S)-4-amino-5-phenylpentanoic acid | 50 g | CAS 1691861-88-7 | InChIKey GQTVSSUIIKDXQC-IBGZPJMESA-N

Catalog No. 502822772
Encompass

Fmoc-(S)-4-amino-5-phenylpentanoic acid is a Amino Acid reagent (Subcategory: Gamma AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1691861-88-7
- MDL: No data
- InChIKey: GQTVSSUIIKDXQC-IBGZPJMESA-N
- Molecular Weight: 415.489
- Molecular Formula: C26H25NO4
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpentanoic acid
- SMILES: O=C(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)N[C@H](CC4=CC=CC=C4)CCC(O)=O

Catalog No. 50-282-2772 Supplier STA Pharmaceutical Supplier No. STAAA1070050GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Fmoc-(S)-4-amino-5-phenylpentanoic acid is a Amino Acid reagent (Subcategory: Gamma AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1691861-88-7
- MDL: No data
- InChIKey: GQTVSSUIIKDXQC-IBGZPJMESA-N
- Molecular Weight: 415.489
- Molecular Formula: C26H25NO4
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpentanoic acid
- SMILES: O=C(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)N[C@H](CC4=CC=CC=C4)CCC(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.