Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC (2S)-3-(1-benzofuran-3-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid | 50 g | InChIKey ASCKURAKVGKPGZ-QHCPKHFHSA-N | MW 427.456

Supplier:  STA PHARMACEUTICAL US LLC STAAA1225150GM

Encompass

(2S)-3-(1-benzofuran-3-yl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid is a Amino Acid reagent (Subcategory: Aryl AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: ASCKURAKVGKPGZ-QHCPKHFHSA-N
- Molecular Weight: 427.456
- Molecular Formula: C26H21NO5
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29329900
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(benzofuran-3-yl)propanoic acid
- SMILES: OC([C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC4=COC5=C4C=CC=C5)=O

Catalog No. 50-282-2056


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.