Learn More
STA PHARMACEUTICAL US LLC Fmoc-beta-L-Phe(2-CN)-OH | 25 g | CAS 270065-84-4 | MDL MFCD01861074
Supplier: STA PHARMACEUTICAL US LLC STAAA1238025GM
Fmoc-beta-L-Phe(2-CN)-OH is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 270065-84-4
- MDL: MFCD01861074
- InChIKey: IIXHWVWQIRQSSX-IBGZPJMESA-N
- Molecular Weight: 426.472
- Molecular Formula: C26H22N2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29269070
- Country Of Origin: China
- IUPAC: (S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(2-cyanophenyl)butanoic acid
- SMILES: OC(C[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC4=C(C#N)C=CC=C4)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.