Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC 4-(Fmoc-amino)-1-(3-Cl-benzyl)-4-carboxypiperidine | 25 g | CAS 916421-89-1 | MDL MFCD08751579

Supplier:  STA PHARMACEUTICAL US LLC STAAA1238425GM

Encompass

4-(Fmoc-amino)-1-(3-Cl-benzyl)-4-carboxypiperidine is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 916421-89-1
- MDL: MFCD08751579
- InChIKey: INMFJSHNRNHCCO-UHFFFAOYSA-N
- Molecular Weight: 490.984
- Molecular Formula: C28H27ClN2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 4-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-1-(3-chlorobenzyl)piperidine-4-carboxylic acid
- SMILES: OC(C1(NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)CCN(CC1)CC5=CC(Cl)=CC=C5)=O

Catalog No. 50-282-1657


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.