Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-Tyr(3-NO2)-OH | 10 g | CAS 181952-16-9 | MDL MFCD15529904

Catalog No. 502822737
Encompass

Fmoc-D-Tyr(3-NO2)-OH is a Amino Acid reagent (Subcategory: Tyr) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 181952-16-9
- MDL: MFCD15529904
- InChIKey: JZUZJVFERQWLNC-HXUWFJFHSA-N
- Molecular Weight: 448.431
- Molecular Formula: C24H20N2O7
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxy-3-nitrophenyl)propanoic acid
- SMILES: OC([C@H](NC(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O)CC4=CC=C(C([N+]([O-])=O)=C4)O)=O

Catalog No. 50-282-2737 Supplier STA Pharmaceutical Supplier No. STAAA1242610GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-Tyr(3-NO2)-OH is a Amino Acid reagent (Subcategory: Tyr) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 181952-16-9
- MDL: MFCD15529904
- InChIKey: JZUZJVFERQWLNC-HXUWFJFHSA-N
- Molecular Weight: 448.431
- Molecular Formula: C24H20N2O7
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxy-3-nitrophenyl)propanoic acid
- SMILES: OC([C@H](NC(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O)CC4=CC=C(C([N+]([O-])=O)=C4)O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.