Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-L-hLys(Alloc)-OH | 100 g | CAS 281655-70-7 | InChIKey KZCWEDOQGDZLTE-QHCPKHFHSA-N

Catalog No. 502822674
Encompass

Fmoc-L-hLys(Alloc)-OH is a Amino Acid reagent (Subcategory: Homo-natural AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 281655-70-7
- MDL: No data
- InChIKey: KZCWEDOQGDZLTE-QHCPKHFHSA-N
- Molecular Weight: 466.534
- Molecular Formula: C26H30N2O6
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-7-(((allyloxy)carbonyl)amino)heptanoic acid
- SMILES: OC([C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CCCCCNC(OCC=C)=O)=O

Catalog No. 50-282-2674 Supplier STA Pharmaceutical Supplier No. STAAA12448100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Fmoc-L-hLys(Alloc)-OH is a Amino Acid reagent (Subcategory: Homo-natural AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 281655-70-7
- MDL: No data
- InChIKey: KZCWEDOQGDZLTE-QHCPKHFHSA-N
- Molecular Weight: 466.534
- Molecular Formula: C26H30N2O6
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-7-(((allyloxy)carbonyl)amino)heptanoic acid
- SMILES: OC([C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CCCCCNC(OCC=C)=O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.