Learn More
STA PHARMACEUTICAL US LLC (R)-2-(Fmoc-amino)-8-azido-2-methyloctanoic acid | 10 g | CAS 1191429-14-7 | MDL MFCD22377534
Supplier: STA PHARMACEUTICAL US LLC STAAA1248910GM
(R)-2-(Fmoc-amino)-8-azido-2-methyloctanoic acid is a Amino Acid reagent (Subcategory: Azido/Alkynyl AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1191429-14-7
- MDL: MFCD22377534
- InChIKey: MSKNYBLJPSFPBR-XMMPIXPASA-N
- Molecular Weight: 436.512
- Molecular Formula: C24H28N4O4
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-8-azido-2-methyloctanoic acid
- SMILES: C[C@](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)(C(O)=O)CCCCCCN=[N+]=[N-]
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.