Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC Fmoc-L-Phe(3F-4COOtBu)-OH | 10 g | InChIKey NJFZOHPQIGMEGS-VWLOTQADSA-N | MW 505.542

Supplier:  STA PHARMACEUTICAL US LLC STAAA1250510GM

Encompass

Fmoc-L-Phe(3F-4COOtBu)-OH is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: NJFZOHPQIGMEGS-VWLOTQADSA-N
- Molecular Weight: 505.542
- Molecular Formula: C29H28FNO6
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxycarbonyl)-3-fluorophenyl)propanoic acid
- SMILES: CC(OC(C1=C(F)C=C(C[C@@H](C(O)=O)NC(OCC(C2=C3C=CC=C2)C4=C3C=CC=C4)=O)C=C1)=O)(C)C

Catalog No. 50-282-1042


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.