Learn More
STA PHARMACEUTICAL US LLC Fmoc-L-Phe(3CHO-4OMe)-OH | 25 g | InChIKey QWAACBANLXKAAA-QHCPKHFHSA-N | MW 445.471
Supplier: STA PHARMACEUTICAL US LLC STAAA1257725GM
Fmoc-L-Phe(3CHO-4OMe)-OH is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: QWAACBANLXKAAA-QHCPKHFHSA-N
- Molecular Weight: 445.471
- Molecular Formula: C26H23NO6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-formyl-4-methoxyphenyl)propanoic acid
- SMILES: COC1=CC=C(C=C1C=O)C[C@@H](C(O)=O)NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.