Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 3-(9H-fluoren-9-ylmethoxycarbonylamino)norbornane-2-carboxylic acid | 100 g | CAS 1217747-47-1 | MDL MFCD20306016

Catalog No. 502820198
Encompass

3-(9H-fluoren-9-ylmethoxycarbonylamino)norbornane-2-carboxylic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1217747-47-1
- MDL: MFCD20306016
- InChIKey: RAQPAPMBITVVOX-UHFFFAOYSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)bicyclo[2.2.1]heptane-2-carboxylic acid
- SMILES: OC(C1C2CCC(C1NC(OCC3C4=C(C5=C3C=CC=C5)C=CC=C4)=O)C2)=O

Catalog No. 50-282-0198 Supplier STA Pharmaceutical Supplier No. STAAA12579100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

3-(9H-fluoren-9-ylmethoxycarbonylamino)norbornane-2-carboxylic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1217747-47-1
- MDL: MFCD20306016
- InChIKey: RAQPAPMBITVVOX-UHFFFAOYSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)bicyclo[2.2.1]heptane-2-carboxylic acid
- SMILES: OC(C1C2CCC(C1NC(OCC3C4=C(C5=C3C=CC=C5)C=CC=C4)=O)C2)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.