Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC BCN-Osu | 10 g | CAS 1426827-79-3 | MDL MFCD19705416

Supplier:  STA PHARMACEUTICAL US LLC STAAA1260910GM

Encompass

BCN-Osu is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1426827-79-3
- MDL: MFCD19705416
- InChIKey: SKTDJYHCSCYLQU-ZSBIGDGJSA-N
- Molecular Weight: 291.303
- Molecular Formula: C15H17NO5
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29280090
- Country Of Origin: China
- IUPAC: ((1R,8S,9s)-bicyclo[6.1.0]non-4-yn-9-yl)methyl (2,5-dioxopyrrolidin-1-yl) carbonate
- SMILES: [H][C@@]12CCC#CCC[C@@]1([C@@H]2COC(ON3C(CCC3=O)=O)=O)[H]

Catalog No. 50-282-0442


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.