Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical N-Fmoc-N-(3,3,3-trifluoropropyl)glycine | 25 g | CAS 2172448-56-3 | InChIKey URYBKKSQSCMAAH-UHFFFAOYSA-N

Catalog No. 502820773
Encompass

N-Fmoc-N-(3,3,3-trifluoropropyl)glycine is a Amino Acid reagent (Subcategory: Gly) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 2172448-56-3
- MDL: No data
- InChIKey: URYBKKSQSCMAAH-UHFFFAOYSA-N
- Molecular Weight: 393.362
- Molecular Formula: C20H18F3NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-(3,3,3-trifluoropropyl)glycine
- SMILES: OC(CN(C(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O)CCC(F)(F)F)=O

Catalog No. 50-282-0773 Supplier STA Pharmaceutical Supplier No. STAAA1263925GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

N-Fmoc-N-(3,3,3-trifluoropropyl)glycine is a Amino Acid reagent (Subcategory: Gly) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 2172448-56-3
- MDL: No data
- InChIKey: URYBKKSQSCMAAH-UHFFFAOYSA-N
- Molecular Weight: 393.362
- Molecular Formula: C20H18F3NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-(3,3,3-trifluoropropyl)glycine
- SMILES: OC(CN(C(OCC1C2=CC=CC=C2C3=CC=CC=C13)=O)CCC(F)(F)F)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.