Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-Lys(Me3)-OH chloride | 10 g | CAS 201004-29-7 | InChIKey XUJRNPVABVHOAJ-FTBISJDPSA-O

Catalog No. 502820573
Encompass

Fmoc-Lys(Me3)-OH chloride is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 201004-29-7
- MDL: No data
- InChIKey: XUJRNPVABVHOAJ-FTBISJDPSA-O
- Molecular Weight: 447.979
- Molecular Formula: C24H32ClN2O4+
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: Unable to generate value
- SMILES: C[N+](C)(CCCC[C@@H](C(O)=O)NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C.Cl

Catalog No. 50-282-0573 Supplier STA Pharmaceutical Supplier No. STAAA1269010GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Lys(Me3)-OH chloride is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 201004-29-7
- MDL: No data
- InChIKey: XUJRNPVABVHOAJ-FTBISJDPSA-O
- Molecular Weight: 447.979
- Molecular Formula: C24H32ClN2O4+
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: Unable to generate value
- SMILES: C[N+](C)(CCCC[C@@H](C(O)=O)NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C.Cl

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.