Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical propargyl-L-lysine hydrochloride | 1 g | CAS 1428330-91-9 | MDL MFCD24842520

Catalog No. 502859246
Encompass

propargyl-L-lysine hydrochloride is a Amino Acid reagent (Subcategory: Azido/Alkynyl AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1428330-91-9
- MDL: MFCD24842520
- InChIKey: ZYUXGVUWYHHZTD-QRPNPIFTSA-N
- Molecular Weight: 264.706
- Molecular Formula: C10H17ClN2O4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29241900
- Country Of Origin: China
- IUPAC: N6-((prop-2-yn-1-yloxy)carbonyl)-L-lysine hydrochloride
- SMILES: N[C@H](C(O)=O)CCCCNC(OCC#C)=O.Cl

Catalog No. 50-285-9246 Supplier STA Pharmaceutical Supplier No. STAAA127331GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

propargyl-L-lysine hydrochloride is a Amino Acid reagent (Subcategory: Azido/Alkynyl AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1428330-91-9
- MDL: MFCD24842520
- InChIKey: ZYUXGVUWYHHZTD-QRPNPIFTSA-N
- Molecular Weight: 264.706
- Molecular Formula: C10H17ClN2O4
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29241900
- Country Of Origin: China
- IUPAC: N6-((prop-2-yn-1-yloxy)carbonyl)-L-lysine hydrochloride
- SMILES: N[C@H](C(O)=O)CCCCNC(OCC#C)=O.Cl

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.