Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Boc-D-N-Me-Orn(Fmoc)-OH | 25 g | CAS 2044710-03-2 | InChIKey CQODNRLCAAIRTG-JOCHJYFZSA-N

Catalog No. 502861102
Encompass

Boc-D-N-Me-Orn(Fmoc)-OH is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 2044710-03-2
- MDL: No data
- InChIKey: CQODNRLCAAIRTG-JOCHJYFZSA-N
- Molecular Weight: 468.55
- Molecular Formula: C26H32N2O6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-5-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)(methyl)amino)pentanoic acid
- SMILES: CN(C(OC(C)(C)C)=O)[C@@H](C(O)=O)CCCNC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O

Catalog No. 50-286-1102 Supplier STA Pharmaceutical Supplier No. STAAA1274125GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Boc-D-N-Me-Orn(Fmoc)-OH is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 2044710-03-2
- MDL: No data
- InChIKey: CQODNRLCAAIRTG-JOCHJYFZSA-N
- Molecular Weight: 468.55
- Molecular Formula: C26H32N2O6
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-5-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)(methyl)amino)pentanoic acid
- SMILES: CN(C(OC(C)(C)C)=O)[C@@H](C(O)=O)CCCNC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.