Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-Lys(diEt)-OH HCl | 25 g | CAS 1451046-67-5 | InChIKey OFODIWHBKXAZFH-BQAIUKQQSA-N

Catalog No. 502859086
Encompass

Fmoc-Lys(diEt)-OH HCl is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1451046-67-5
- MDL: No data
- InChIKey: OFODIWHBKXAZFH-BQAIUKQQSA-N
- Molecular Weight: 460.999
- Molecular Formula: C25H33ClN2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6,N6-diethyl-L-lysine hydrochloride
- SMILES: CCN(CCCC[C@@H](C(O)=O)NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC.Cl

Catalog No. 50-285-9086 Supplier STA Pharmaceutical Supplier No. STAAA1280125GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Fmoc-Lys(diEt)-OH HCl is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1451046-67-5
- MDL: No data
- InChIKey: OFODIWHBKXAZFH-BQAIUKQQSA-N
- Molecular Weight: 460.999
- Molecular Formula: C25H33ClN2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N2-(((9H-fluoren-9-yl)methoxy)carbonyl)-N6,N6-diethyl-L-lysine hydrochloride
- SMILES: CCN(CCCC[C@@H](C(O)=O)NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)CC.Cl

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.