Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical LNA A(pac) amidite | 200 mg | InChIKey RHUYJRYFCPQBIE-LXTWGDFWSA-N | MW 915.984761998

Catalog No. 502859723
Encompass

LNA A(pac) amidite is a Amidite reagent (Subcategory: LNA Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: RHUYJRYFCPQBIE-LXTWGDFWSA-N
- Molecular Weight: 915.984761998
- Molecular Formula: C49H54N7O9P
- Purity: ≥97%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (1R,3R,4R,7S)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3-(6-(2-phenoxyacetamido)-9H-purin-9-yl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]2OC[C@]1(COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)O[C@H]2N6C7=C(C(NC(COC8=CC=CC=C8)=O)=NC=N7)N=C6)OCCC#N)C(C)C)C

Catalog No. 50-285-9723 Supplier STA Pharmaceutical Supplier No. STAAM10203200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LNA A(pac) amidite is a Amidite reagent (Subcategory: LNA Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: RHUYJRYFCPQBIE-LXTWGDFWSA-N
- Molecular Weight: 915.984761998
- Molecular Formula: C49H54N7O9P
- Purity: ≥97%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (1R,3R,4R,7S)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-3-(6-(2-phenoxyacetamido)-9H-purin-9-yl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]2OC[C@]1(COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)O[C@H]2N6C7=C(C(NC(COC8=CC=CC=C8)=O)=NC=N7)N=C6)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.