Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 5’-Biotin C6 amine amidite | 1 g | CAS 135137-87-0 | InChIKey RBXYKQBFPLXSJS-UHFFFAOYSA-N

Catalog No. 502859328
Encompass

5’-Biotin C6 amine amidite is a Amidite reagent (Subcategory: Specialty Linker/End Modifier) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 135137-87-0
- MDL: No data
- InChIKey: RBXYKQBFPLXSJS-UHFFFAOYSA-N
- Molecular Weight: 846.080761998
- Molecular Formula: C46H64N5O6PS
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 6-(5-(1-(bis(4-methoxyphenyl)(phenyl)methyl)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C(NCCCCCCOP(N(C(C)C)C(C)C)OCCC#N)CCCCC1SCC(C1NC2=O)N2C(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5

Catalog No. 50-285-9328 Supplier STA Pharmaceutical Supplier No. STAAM10336AK601GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5’-Biotin C6 amine amidite is a Amidite reagent (Subcategory: Specialty Linker/End Modifier) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 135137-87-0
- MDL: No data
- InChIKey: RBXYKQBFPLXSJS-UHFFFAOYSA-N
- Molecular Weight: 846.080761998
- Molecular Formula: C46H64N5O6PS
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 6-(5-(1-(bis(4-methoxyphenyl)(phenyl)methyl)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C(NCCCCCCOP(N(C(C)C)C(C)C)OCCC#N)CCCCC1SCC(C1NC2=O)N2C(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.