Learn More
STA PHARMACEUTICAL US LLC aTNA MeC(Bz) amidite | 1 g | InChIKey LBIIXWXTPKSQDV-ORHIHEHSSA-N | MW 876.991761998
Supplier: STA PHARMACEUTICAL US LLC STAAM10393AK601GM
aTNA MeC(Bz) amidite is a Amidite reagent (Subcategory: aTNA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: LBIIXWXTPKSQDV-ORHIHEHSSA-N
- Molecular Weight: 876.991761998
- Molecular Formula: C48H57N6O8P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: (2R,3R)-3-(2-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)acetamido)-4-(bis(4-methoxyphenyl)(phenyl)methoxy)butan-2-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: C[C@@H](OP(N(C(C)C)C(C)C)OCCC#N)[C@H](NC(CN1C(N=C(NC(C2=CC=CC=C2)=O)C(C)=C1)=O)=O)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.