Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical PMO A(Bz) Monomer | 25 g | CAS 956139-18-7 | InChIKey HRNHOEBQNIMEGD-RJFDGRJRSA-N

Catalog No. 502860565
Encompass

PMO A(Bz) Monomer is a Amidite reagent (Subcategory: PMO Monomer) sold by WuXi TIDES. Offered in 25 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 956139-18-7
- MDL: No data
- InChIKey: HRNHOEBQNIMEGD-RJFDGRJRSA-N
- Molecular Weight: 722.182761998
- Molecular Formula: C38H37ClN7O4P
- Purity: ≥99%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: ((2S,6R)-6-(6-benzamido-9H-purin-9-yl)-4-tritylmorpholin-2-yl)methyl dimethylphosphoramidochloridate
- SMILES: O=P(Cl)(OC[C@H]1O[C@H](CN(C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)N5C6=C(N=C5)C(NC(C7=CC=CC=C7)=O)=NC=N6)N(C)C

Catalog No. 50-286-0565 Supplier STA Pharmaceutical Supplier No. STAAM1057225GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PMO A(Bz) Monomer is a Amidite reagent (Subcategory: PMO Monomer) sold by WuXi TIDES. Offered in 25 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 956139-18-7
- MDL: No data
- InChIKey: HRNHOEBQNIMEGD-RJFDGRJRSA-N
- Molecular Weight: 722.182761998
- Molecular Formula: C38H37ClN7O4P
- Purity: ≥99%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: ((2S,6R)-6-(6-benzamido-9H-purin-9-yl)-4-tritylmorpholin-2-yl)methyl dimethylphosphoramidochloridate
- SMILES: O=P(Cl)(OC[C@H]1O[C@H](CN(C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)N5C6=C(N=C5)C(NC(C7=CC=CC=C7)=O)=NC=N6)N(C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.