Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical PMO C(Bz) Monomer | 1 g | CAS 956139-21-2 | InChIKey RCQVHNTXMQCQLT-AUIRFOQBSA-N

Catalog No. 502860904
Encompass

PMO C(Bz) Monomer is a Amidite reagent (Subcategory: PMO Monomer) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 956139-21-2
- MDL: No data
- InChIKey: RCQVHNTXMQCQLT-AUIRFOQBSA-N
- Molecular Weight: 698.156761998
- Molecular Formula: C37H37ClN5O5P
- Purity: ≥99%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: ((2S,6R)-6-(4-benzamido-2-oxopyrimidin-1(2H)-yl)-4-tritylmorpholin-2-yl)methyl dimethylphosphoramidochloridate
- SMILES: O=C1N=C(C=CN1[C@H]2CN(C[C@H](O2)COP(N(C)C)(Cl)=O)C(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)NC(C6=CC=CC=C6)=O

Catalog No. 50-286-0904 Supplier STA Pharmaceutical Supplier No. STAAM10574MM301GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PMO C(Bz) Monomer is a Amidite reagent (Subcategory: PMO Monomer) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 956139-21-2
- MDL: No data
- InChIKey: RCQVHNTXMQCQLT-AUIRFOQBSA-N
- Molecular Weight: 698.156761998
- Molecular Formula: C37H37ClN5O5P
- Purity: ≥99%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: ((2S,6R)-6-(4-benzamido-2-oxopyrimidin-1(2H)-yl)-4-tritylmorpholin-2-yl)methyl dimethylphosphoramidochloridate
- SMILES: O=C1N=C(C=CN1[C@H]2CN(C[C@H](O2)COP(N(C)C)(Cl)=O)C(C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5)NC(C6=CC=CC=C6)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.