Learn More
STA PHARMACEUTICAL US LLC 2'-F G(iBu) OMe-amidite | 5 g | InChIKey WEPASXLNAQHNEK-NNYLMNAYSA-N | MW 818.884165161
Supplier: STA PHARMACEUTICAL US LLC STAAM107485GM
2'-F G(iBu) OMe-amidite is a Amidite reagent (Subcategory: OMe-P Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: WEPASXLNAQHNEK-NNYLMNAYSA-N
- Molecular Weight: 818.884165161
- Molecular Formula: C42H52FN6O8P
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-fluoro-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)tetrahydrofuran-3-yl methyl diisopropylphosphoramidite
- SMILES: O=C(C(C)C)NC(N1)=NC2=C(N=CN2[C@@H]3O[C@H](COC(C4=CC=C(C=C4)OC)(C5=CC=C(C=C5)OC)C6=CC=CC=C6)[C@@H](OP(OC)N(C(C)C)C(C)C)[C@H]3F)C1=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.