Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical ANA 2'-OMe G(iBu) amidite | 200 mg | InChIKey IRRDHRZUOZNWDJ-HHNCBLOXSA-N | MW 869.956761998

Catalog No. 502860879
Encompass

ANA 2'-OMe G(iBu) amidite is a Amidite reagent (Subcategory: ANA Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: IRRDHRZUOZNWDJ-HHNCBLOXSA-N
- Molecular Weight: 869.956761998
- Molecular Formula: C45H56N7O9P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1C(N=CN2[C@@H]3O[C@@H]([C@H]([C@@H]3OC)OP(N(C(C)C)C(C)C)OCCC#N)COC(C4=CC=C(C=C4)OC)(C5=CC=C(C=C5)OC)C6=CC=CC=C6)=C2N=C(N1)NC(C(C)C)=O

Catalog No. 50-286-0879 Supplier STA Pharmaceutical Supplier No. STAAM10776200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ANA 2'-OMe G(iBu) amidite is a Amidite reagent (Subcategory: ANA Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: IRRDHRZUOZNWDJ-HHNCBLOXSA-N
- Molecular Weight: 869.956761998
- Molecular Formula: C45H56N7O9P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2-isobutyramido-6-oxo-1,6-dihydro-9H-purin-9-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1C(N=CN2[C@@H]3O[C@@H]([C@H]([C@@H]3OC)OP(N(C(C)C)C(C)C)OCCC#N)COC(C4=CC=C(C=C4)OC)(C5=CC=C(C=C5)OC)C6=CC=CC=C6)=C2N=C(N1)NC(C(C)C)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.