Learn More
STA PHARMACEUTICAL US LLC 2'-OMe C(Ac) OBn-amidite | 1 g | InChIKey LSZOOEVRGCMGII-YDFRAEGJSA-N | MW 838.938761998
Supplier: STA PHARMACEUTICAL US LLC STAAM10787MM301GM
2'-OMe C(Ac) OBn-amidite is a Amidite reagent (Subcategory: OBn-P Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: LSZOOEVRGCMGII-YDFRAEGJSA-N
- Molecular Weight: 838.938761998
- Molecular Formula: C46H55N4O9P
- Purity: ≥95%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-5-(4-acetamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl benzyl diisopropylphosphoramidite
- SMILES: COC1=CC=C(C=C1)C(OC[C@H]2O[C@H]([C@@H]([C@@H]2OP(N(C(C)C)C(C)C)OCC3=CC=CC=C3)OC)N4C=CC(NC(C)=O)=NC4=O)(C5=CC=CC=C5)C6=CC=C(C=C6)OC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.