Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical DNA 5-I-C(dbf) amidite | 1 g | InChIKey XQOVLOSUWOGEKK-XHIGJUGSSA-N | MW 994.953231998

Catalog No. 502860882
Encompass

DNA 5-I-C(dbf) amidite is a Amidite reagent (Subcategory: Cytosine Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: XQOVLOSUWOGEKK-XHIGJUGSSA-N
- Molecular Weight: 994.953231998
- Molecular Formula: C48H64IN6O7P
- Purity: ≥95%
- Container Type: 30 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 47.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(4-(((E)-(dibutylamino)methylene)amino)-5-iodo-2-oxopyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N=C(C(I)=CN1[C@H](O[C@@H]2COC(C3=CC=CC=C3)(C4=CC=C(C=C4)OC)C5=CC=C(C=C5)OC)C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)/N=C/N(CCCC)CCCC

Catalog No. 50-286-0882 Supplier STA Pharmaceutical Supplier No. STAAM10792KA301GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA 5-I-C(dbf) amidite is a Amidite reagent (Subcategory: Cytosine Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: XQOVLOSUWOGEKK-XHIGJUGSSA-N
- Molecular Weight: 994.953231998
- Molecular Formula: C48H64IN6O7P
- Purity: ≥95%
- Container Type: 30 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 47.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(4-(((E)-(dibutylamino)methylene)amino)-5-iodo-2-oxopyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N=C(C(I)=CN1[C@H](O[C@@H]2COC(C3=CC=CC=C3)(C4=CC=C(C=C4)OC)C5=CC=C(C=C5)OC)C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)/N=C/N(CCCC)CCCC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.