Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Thio(Ac)-PEG3 amidite | 1 g | CAS 153870-21-4 | InChIKey FMIVUFGXNQLSTH-UHFFFAOYSA-N

Catalog No. 502859588
Encompass

Thio(Ac)-PEG3 amidite is a Amidite reagent (Subcategory: Thiol Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 153870-21-4
- MDL: No data
- InChIKey: FMIVUFGXNQLSTH-UHFFFAOYSA-N
- Molecular Weight: 408.493761998
- Molecular Formula: C17H33N2O5PS
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29309095
- Country Of Origin: China
- IUPAC: S-(2-(2-(2-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)ethoxy)ethoxy)ethyl) ethanethioate
- SMILES: CC(N(P(OCCOCCOCCSC(C)=O)OCCC#N)C(C)C)C

Catalog No. 50-285-9588 Supplier STA Pharmaceutical Supplier No. STAAM108151GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thio(Ac)-PEG3 amidite is a Amidite reagent (Subcategory: Thiol Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 153870-21-4
- MDL: No data
- InChIKey: FMIVUFGXNQLSTH-UHFFFAOYSA-N
- Molecular Weight: 408.493761998
- Molecular Formula: C17H33N2O5PS
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29309095
- Country Of Origin: China
- IUPAC: S-(2-(2-(2-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)ethoxy)ethoxy)ethyl) ethanethioate
- SMILES: CC(N(P(OCCOCCOCCSC(C)=O)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.