Learn More
STA PHARMACEUTICAL US LLC O1-DMTr-O3-[3'-(Cholesterol)amino-Propyl-PEG3]-Glycerin amidite | 5 g | CAS 873435-29-1 | InChIKey FNJSTUYYFCEZCA-PORNTWEGSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10827AK1005GM
O1-DMTr-O3-[3'-(Cholesterol)amino-Propyl-PEG3]-Glycerin amidite is a Amidite reagent (Subcategory: Cholesterol Modifier) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 873435-29-1
- MDL: No data
- InChIKey: FNJSTUYYFCEZCA-PORNTWEGSA-N
- Molecular Weight: 1196.601761998
- Molecular Formula: C70H106N3O11P
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29299090
- Country Of Origin: China
- SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@]2([H])[C@]1(C)CC[C@@]3([H])[C@@]2([H])CC=C4[C@]3(C)CC[C@H](OC(NCCCOCCOCCOCCOCC(COC(C5=CC=C(OC)C=C5)(C6=CC=CC=C6)C7=CC=C(OC)C=C7)OP(N(C(C)C)C(C)C)OCCC#N)=O)C4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.