Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 5'-Hexynyl C6 amidite | 5 g | CAS 1048985-37-0 | InChIKey IFNKHZIJWLUIHZ-UHFFFAOYSA-N

Catalog No. 502858489
Encompass

5'-Hexynyl C6 amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 1048985-37-0
- MDL: No data
- InChIKey: IFNKHZIJWLUIHZ-UHFFFAOYSA-N
- Molecular Weight: 298.366761998
- Molecular Formula: C15H27N2O2P
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl hex-5-yn-1-yl diisopropylphosphoramidite
- SMILES: C#CCCCCOP(OCCC#N)N(C(C)C)C(C)C

Catalog No. 50-285-8489 Supplier STA Pharmaceutical Supplier No. STAAM108465GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-Hexynyl C6 amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 1048985-37-0
- MDL: No data
- InChIKey: IFNKHZIJWLUIHZ-UHFFFAOYSA-N
- Molecular Weight: 298.366761998
- Molecular Formula: C15H27N2O2P
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl hex-5-yn-1-yl diisopropylphosphoramidite
- SMILES: C#CCCCCOP(OCCC#N)N(C(C)C)C(C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.