Learn More
STA PHARMACEUTICAL US LLC 5'-Hexynyl C6 amidite | 200 mg | CAS 1048985-37-0 | InChIKey IFNKHZIJWLUIHZ-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10846200MG
5'-Hexynyl C6 amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 1048985-37-0
- MDL: No data
- InChIKey: IFNKHZIJWLUIHZ-UHFFFAOYSA-N
- Molecular Weight: 298.366761998
- Molecular Formula: C15H27N2O2P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl hex-5-yn-1-yl diisopropylphosphoramidite
- SMILES: C#CCCCCOP(OCCC#N)N(C(C)C)C(C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.