Learn More
STA PHARMACEUTICAL US LLC 2'-OMe N3-Bz-U amidite | 1 g | CAS 178422-03-2 | InChIKey RLZUETJITSYKAT-VKBHKTMGSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10876AK601GM
2'-OMe N3-Bz-U amidite is a Amidite reagent (Subcategory: Uracil Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 178422-03-2
- MDL: No data
- InChIKey: RLZUETJITSYKAT-VKBHKTMGSA-N
- Molecular Weight: 864.932761998
- Molecular Formula: C47H53N4O10P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-5-(3-benzoyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(C2=CC=C(OC)C=C2)(C3=CC=CC=C3)C4=CC=C(OC)C=C4)O[C@H]1N5C=CC(N(C5=O)C(C6=CC=CC=C6)=O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.