Learn More
STA PHARMACEUTICAL US LLC DNA T OTBSE-amidite | 5 g | CAS 173440-40-9 | InChIKey OPKCOYPGFFHUTD-STPYIPAVSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10884MM2505GM
DNA T OTBSE-amidite is a Amidite reagent (Subcategory: Silylalkoxy-P Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 173440-40-9
- MDL: No data
- InChIKey: OPKCOYPGFFHUTD-STPYIPAVSA-N
- Molecular Weight: 834.078761998
- Molecular Formula: C45H64N3O8PSi
- Purity: ≥95%
- Container Type: 250 mL Glass (28-400)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 174.7 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-(tert-butyldimethylsilyl)ethyl) diisopropylphosphoramidite
- SMILES: O=C(NC1=O)N([C@@H]2O[C@H](COC(C3=CC=C(C=C3)OC)(C4=CC=CC=C4)C5=CC=C(C=C5)OC)[C@@H](OP(N(C(C)C)C(C)C)OCC[Si](C)(C(C)(C)C)C)C2)C=C1C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.