Learn More
STA PHARMACEUTICAL US LLC TFA C3 amine amidite | 5 g | CAS 853955-89-2 | InChIKey AOAXRHUQUZSDAX-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10962AK1005GM
TFA C3 amine amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 853955-89-2
- MDL: No data
- InChIKey: AOAXRHUQUZSDAX-UHFFFAOYSA-N
- Molecular Weight: 371.340971487
- Molecular Formula: C14H25F3N3O3P
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (3-(2,2,2-trifluoroacetamido)propyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OCCCNC(C(F)(F)F)=O)OCCC#N)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.