Learn More
STA PHARMACEUTICAL US LLC 2'-TOM G(iPr-pac) amidite | 200 mg | InChIKey MCJWYJDVECOECQ-VPEMNPORSA-N | MW 1148.423761998
Supplier: STA PHARMACEUTICAL US LLC STAAM11009200MG
2'-TOM G(iPr-pac) amidite is a Amidite reagent (Subcategory: 2'-OH Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: MCJWYJDVECOECQ-VPEMNPORSA-N
- Molecular Weight: 1148.423761998
- Molecular Formula: C61H82N7O11PSi
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2-(2-(4-isopropylphenoxy)acetamido)-6-oxo-1,6-dihydro-9H-purin-9-yl)-4-(((triisopropylsilyl)oxy)methoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: COC1=CC=C(C=C1)C(OC[C@@H]2[C@H]([C@H]([C@@H](O2)N3C=NC4=C3N=C(NC4=O)NC(COC5=CC=C(C=C5)C(C)C)=O)OCO[Si](C(C)C)(C(C)C)C(C)C)OP(OCCC#N)N(C(C)C)C(C)C)(C6=CC=C(C=C6)OC)C7=CC=CC=C7
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.