Learn More
STA PHARMACEUTICAL US LLC DNA MeC(Bz) PACE amidite | 5 g | InChIKey SCWQJBSKQDMVJI-PTJYUUIYSA-N | MW 918.040761998
Supplier: STA PHARMACEUTICAL US LLC STAAM11106AK1005GM
DNA MeC(Bz) PACE amidite is a Amidite reagent (Subcategory: PACE Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.
Specifications
- CAS: No data
- MDL: No data
- InChIKey: SCWQJBSKQDMVJI-PTJYUUIYSA-N
- Molecular Weight: 918.040761998
- Molecular Formula: C51H60N5O9P
- Purity: ≥95%
- Container Type: 100 mL Glass (Septum)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 100.4 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 1-cyano-2-methylpropan-2-yl 2-((((2R,3S,5R)-5-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl)oxy)(diisopropylamino)phosphaneyl)acetate
- SMILES: CC(OC(CP(N(C(C)C)C(C)C)O[C@@H]1[C@@H](COC(C2=CC=C(OC)C=C2)(C3=CC=CC=C3)C4=CC=C(OC)C=C4)O[C@@H](N5C=C(C)C(NC(C6=CC=CC=C6)=O)=NC5=O)C1)=O)(CC#N)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.