Learn More
DIAGNOSTICA STAGO INC STA-Coag Control N ABN Plus, Comprehensive Coagulation Control, 24-Hour Stability, 2 mL/vial
Catalog No. NC1578116
Encompass
The STA-Coag Control N ABN Plus is a comprehensive coagulation control plasma designed for quality control in hemostasis testing. It provides both normal and abnormal control levels, ensuring accurate assay performance across various coagulation parameters. With broad analyte coverage and 24-hour onboard stability after reconstitution, it minimizes waste, enhances laboratory efficiency, and supports reliable diagnostic results, making it an essential tool for laboratories.
This item is not returnable. View return policy
Catalog No. NC1578116 Supplier DIAGNOSTICA STAGO INC Supplier No. 00677
The STA-Coag Control N ABN Plus is a comprehensive coagulation control plasma designed for quality control in hemostasis testing. It provides both normal and abnormal control levels, ensuring accurate assay performance across various coagulation parameters. With broad analyte coverage and 24-hour onboard stability after reconstitution, it minimizes waste, enhances laboratory efficiency, and supports reliable diagnostic results, making it an essential tool for laboratories.