Learn More
Santa Cruz Biotechnology VSV-G Antibody (F-6) | 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC365019
Encompass
Anti-VSV-G Antibody (F-6) is a mouse monoclonal IgG2b κ VSV-G antibody, cited in 6 publications, provided at 200 µg/ml. Specific for an epitope mapping between amino acids 493-511 at the C-terminus of VSV-G of Vesicular Stomatitis virus origin. VSV-G Antibody (F-6) is recommended for detection of VSV-G of VSV origin by WB, IP, IF and ELISA. Anti-VSV-G Antibody (F-6) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM.
Catalog No. NC1760772