Learn More
MSC Stabila Measuring Stick, 48 in, 3 Vial, Aluminum
Catalog No. 5000528269
Encompass
This measuring stick is designed for professional remodeling, reconstruction, and restoration, making kitchen, bath, closet, or basement remodels faster and easier.
  • Plumb and level with three measuring scales on the frame.
  • Level and mark at the same time.
  • Red rubber pads on both sides engage the wall without slipping or causing scratches.
  • Straight-edge frame allows for uninterrupted scribing, scoring, and tracing.
  • Accuracy is guaranteed for life; vials will never fog, leak, or require adjustment.
  • The frame is easy to grip, read, and clean, with no hand grips or ribs.
  • Six ring vials read level plus 1% and 2% slope.
  • Features two plumb and one level vial.
  • Smooth rectangular profile.
Catalog No. 50-005-28269 Supplier MSC Supplier No. 37426921
This measuring stick is designed for professional remodeling, reconstruction, and restoration, making kitchen, bath, closet, or basement remodels faster and easier.
  • Plumb and level with three measuring scales on the frame.
  • Level and mark at the same time.
  • Red rubber pads on both sides engage the wall without slipping or causing scratches.
  • Straight-edge frame allows for uninterrupted scribing, scoring, and tracing.
  • Accuracy is guaranteed for life; vials will never fog, leak, or require adjustment.
  • The frame is easy to grip, read, and clean, with no hand grips or ribs.
  • Six ring vials read level plus 1% and 2% slope.
  • Features two plumb and one level vial.
  • Smooth rectangular profile.