Learn More
Surmodics Inc StabilBlock Immunoassay Stabilizer, 1L
SDP
Catalog No. NC1576428
Encompass_Preferred
StabilBlock Immunoassay Stabilizer effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. It can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper. The key benefits are reduction of non-specific binding while increasing signal-to-noise ratios within immunoassay applications.
Catalog No. NC1576428 Supplier Surmodics Inc Supplier No. ST011000
StabilBlock Immunoassay Stabilizer effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. It can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper. The key benefits are reduction of non-specific binding while increasing signal-to-noise ratios within immunoassay applications.