Learn More
Surmodics Inc StabilBlock Immunoassay Stabilizer 50ml
SDP
Catalog No. NC1213102
Encompass_Preferred
StabilBlock™ Immunoassay Stabilizer effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. It can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper. The key benefits are reduction of non-specific binding while increasing signal-to-noise ratios within immunoassay applications.
Catalog No. NC1213102 Supplier Surmodics Inc Supplier No. ST010050
StabilBlock™ Immunoassay Stabilizer effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. It can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper. The key benefits are reduction of non-specific binding while increasing signal-to-noise ratios within immunoassay applications.