Learn More
Caron Products And Services Stability Freezer, 33 cu. ft., Temp
Catalog No. NC2676689
Encompass_Preferred
Stability Refrigerator/Freezer. 33 cubic foot with temperature control, touchscreen. Includes Automatic defrost. -25C to 10C, 208/230V, 60 Hz.
Catalog No. NC2676689 Supplier Caron Products And Services Supplier No. 7004332
Add to Cart
Add to Cart
Request a Quote
Stability Refrigerator/Freezer. 33 cubic foot with temperature control, touchscreen. Includes Automatic defrost. -25C to 10C, 208/230V, 60 Hz.