Learn More
Genscript Corporation Stabilized dNTP Mix 10 mM eac
Catalog No. 5000058959
Encompass
Suitable for use in PCR sequencing nick translation fill-in cDNA synthesis TdT tailing reactions and dilution of radiolabeled dNTPs These nucleotides are supplied as convenient ready-to-use solutions in Tris-HCl (pH 7 5) The 10 mM HS dNTP Mix contains all four dNTPs each at a final concentration of 10 mM It also contains a stabilizer which makes it a very long-lasting tool
Catalog No. 50-000-58959 Supplier Genscript Corporation Supplier No. C01689
Suitable for use in PCR sequencing nick translation fill-in cDNA synthesis TdT tailing reactions and dilution of radiolabeled dNTPs These nucleotides are supplied as convenient ready-to-use solutions in Tris-HCl (pH 7 5) The 10 mM HS dNTP Mix contains all four dNTPs each at a final concentration of 10 mM It also contains a stabilizer which makes it a very long-lasting tool